The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178098.1 | NP_001171569.1 | 8 - 9 | 1261 | 103 |
NM_001770.5 | NP_001761.3 | 8 - 9 | 1261 | 103 | |
XM_006721103.3 | XP_006721166.1 | 7 - 8 | 1198 | 103 | |
XM_017023893.1 | XP_016879382.1 | 7 - 8 | 1062 | 103 | |
GenBank mRNA | AK130657.1 | - | 7 - 8 | 647 | 103 |
AK301167.1 | - | 8 - 9 | 1179 | 103 | |
AK304456.1 | - | 9 - 10 | 1046 | 103 | |
AK313577.1 | - | 8 - 9 | 1232 | 103 | |
BC006338.2 | - | 8 - 9 | 1243 | 103 | |
M21097.1 | - | 8 - 9 | 1214 | 102 | |
X13312.1 | - | 8 - 9 | 1213 | 102 |