The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135673.2 | NP_001129145.1 | 8 - 9 | 967 | 105 |
NM_001308076.1 | NP_001295005.1 | 8 - 9 | 1234 | 105 | |
NM_022374.3 | NP_071769.2 | 8 - 9 | 967 | 105 | |
NR_024191.1 | 7 - 8 | 722 | 105 | ||
XM_011533017.1 | XP_011531319.1 | 8 - 9 | 967 | 105 | |
XM_011533018.2 | XP_011531320.1 | 8 - 9 | 1377 | 105 | |
XM_011533019.2 | XP_011531321.1 | 8 - 9 | 1385 | 105 | |
XM_011533020.1 | XP_011531322.1 | 8 - 9 | 1065 | 105 | |
XM_011533022.2 | XP_011531324.1 | 7 - 8 | 717 | 105 | |
XM_011533023.2 | XP_011531325.1 | 8 - 9 | 2252 | 105 | |
XM_017004686.1 | XP_016860175.1 | 8 - 9 | 930 | 105 | |
XM_017004687.1 | XP_016860176.1 | 8 - 9 | 2112 | 105 | |
XM_017004688.1 | XP_016860177.1 | 8 - 9 | 2252 | 105 | |
GenBank mRNA | AK026946.1 | - | 8 - 9 | 950 | 105 |
AK225190.1 | - | 8 - 9 | 953 | 105 | |
AK294049.1 | - | 8 - 9 | 928 | 105 | |
AK294711.1 | - | 7 - 8 | 683 | 105 | |
AK302621.1 | - | 8 - 9 | 1234 | 105 | |
AK303985.1 | - | 7 - 8 | 706 | 105 | |
AK308894.1 | - | 7 - 8 | 707 | 105 | |
BC053508.1 | - | 7 - 8 | 694 | 105 |