Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOXA1
Entrez Gene ID
10811
Gene Name
NADPH oxidase activator 1
Gene Aliases
NY-CO-31, SDCCAG31, p51NOX
Chromosome Location
Chr.9: 137423372 - 137434406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256067.1 NP_001242996.1 13 - 14 1432 120
GenBank mRNA AB095031.1 - 13 - 14 1412 120
AK098341.1 - 11 - 12 1357 120
AY255769.1 - 13 - 14 1297 120
AY927790.1 - 13 - 14 1335 120

View Full Product Details