The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030581.3 | NP_085058.3 | 23 - 24 | 2520 | 93 |
XM_005256146.3 | XP_005256203.1 | 24 - 25 | 2581 | 93 | |
XM_011523331.2 | XP_011521633.1 | 22 - 23 | 2341 | 93 | |
XM_011523332.2 | XP_011521634.1 | 18 - 19 | 2029 | 93 | |
XM_017023667.1 | XP_016879156.1 | 22 - 23 | 2381 | 93 | |
XM_017023668.1 | XP_016879157.1 | 21 - 22 | 2157 | 93 | |
XM_017023669.1 | XP_016879158.1 | 22 - 23 | 2229 | 93 | |
XM_017023670.1 | XP_016879159.1 | 21 - 22 | 2262 | 93 | |
GenBank mRNA | AB067510.1 | - | 18 - 19 | 1993 | 93 |
AF370390.1 | - | 11 - 12 | 1452 | 93 | |
AK022332.1 | - | 6 - 7 | 952 | 93 | |
AK091316.1 | - | 11 - 12 | 1300 | 93 | |
AL832078.1 | - | 8 - 9 | 928 | 93 | |
BC014887.2 | - | 17 - 18 | 1912 | 93 |