The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324171.1 | NP_001311100.1 | 7 - 8 | 659 | 60 |
NM_001324172.1 | NP_001311101.1 | 7 - 8 | 659 | 60 | |
NM_030581.3 | NP_085058.3 | 7 - 8 | 632 | 60 | |
XM_005256146.3 | XP_005256203.1 | 7 - 8 | 636 | 60 | |
XM_011523331.2 | XP_011521633.1 | 6 - 7 | 476 | 60 | |
XM_011523332.2 | XP_011521634.1 | 1 - 2 | 59 | 60 | |
XM_017023669.1 | XP_016879158.1 | 6 - 7 | 364 | 60 | |
XM_017023670.1 | XP_016879159.1 | 5 - 6 | 372 | 60 | |
GenBank mRNA | AB067510.1 | - | 2 - 3 | 105 | 60 |
AK302557.1 | - | 7 - 8 | 700 | 60 | |
BC004519.2 | - | 7 - 8 | 596 | 60 | |
BU850134.1 | - | 7 - 8 | 584 | 60 |