Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
Chromosome Location
Chr.16: 74873568 - 74985146 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324171.1 NP_001311100.1 6 - 7 574 82
NM_001324172.1 NP_001311101.1 6 - 7 574 82
NM_030581.3 NP_085058.3 6 - 7 547 82
XM_005256146.3 XP_005256203.1 6 - 7 551 82
XM_011523331.2 XP_011521633.1 5 - 6 391 82
XM_017023667.1 XP_016879156.1 5 - 6 391 82
XM_017023669.1 XP_016879158.1 5 - 6 279 82
XM_017023670.1 XP_016879159.1 4 - 5 287 82
GenBank mRNA AK302557.1 - 6 - 7 615 82
BC004519.2 - 6 - 7 511 82
BU850134.1 - 6 - 7 499 82

View Full Product Details