The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014729.2 | NP_055544.1 | 4 - 5 | 915 | 118 |
XM_017014085.1 | XP_016869574.1 | 3 - 4 | 934 | 118 | |
GenBank mRNA | AB018351.1 | - | 4 - 5 | 917 | 118 |
BC016665.1 | - | 4 - 5 | 758 | 118 |