The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322124.1 | NP_001309053.1 | 1 - 2 | 58 | 78 |
GenBank mRNA | AW503429.1 | - | 1 - 2 | 65 | 78 |