The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322123.1 | NP_001309052.1 | 10 - 11 | 1082 | 103 |
NM_001322124.1 | NP_001309053.1 | 10 - 11 | 951 | 103 | |
NM_001322127.1 | NP_001309056.1 | 9 - 10 | 1077 | 103 | |
NM_144709.3 | NP_653310.2 | 10 - 11 | 1139 | 103 | |
XM_011532568.2 | XP_011530870.1 | 11 - 12 | 1210 | 103 | |
XM_011532570.1 | XP_011530872.1 | 10 - 11 | 917 | 103 | |
XM_011532571.1 | XP_011530873.1 | 10 - 11 | 1091 | 103 | |
XM_011532572.1 | XP_011530874.1 | 10 - 11 | 1011 | 103 | |
XM_011532573.2 | XP_011530875.1 | 10 - 11 | 971 | 103 | |
XM_011532574.2 | XP_011530876.1 | 10 - 11 | 1032 | 103 | |
XM_011532576.2 | XP_011530878.1 | 10 - 11 | 1091 | 103 | |
XM_011532578.1 | XP_011530880.1 | 10 - 11 | 1069 | 103 | |
XM_017003428.1 | XP_016858917.1 | 9 - 10 | 908 | 103 | |
XM_017003429.1 | XP_016858918.1 | 9 - 10 | 704 | 103 | |
GenBank mRNA | AK056874.1 | - | 10 - 11 | 953 | 103 |
AK291729.1 | - | 10 - 11 | 927 | 103 | |
AL832208.1 | - | 8 - 9 | 3478 | 103 | |
BC101680.1 | - | 10 - 11 | 931 | 103 | |
BC101706.1 | - | 10 - 11 | 931 | 103 | |
BC143480.1 | - | 9 - 10 | 869 | 103 |