The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322127.1 | NP_001309056.1 | 1 - 2 | 251 | 83 |
NM_144709.3 | NP_653310.2 | 1 - 2 | 251 | 83 | |
XM_017003428.1 | XP_016858917.1 | 1 - 2 | 275 | 83 | |
GenBank mRNA | AK056874.1 | - | 1 - 2 | 65 | 83 |
AL832208.1 | - | 1 - 2 | 252 | 83 | |
BC101680.1 | - | 1 - 2 | 43 | 83 | |
BC101706.1 | - | 1 - 2 | 43 | 83 | |
BC143480.1 | - | 1 - 2 | 43 | 83 | |
BP354604.1 | - | 1 - 2 | 231 | 83 |