The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134225.1 | NP_001127697.1 | 13 - 14 | 1559 | 89 |
XM_006712501.3 | XP_006712564.1 | 13 - 14 | 1554 | 89 | |
XM_011511124.2 | XP_011509426.1 | 13 - 14 | 1554 | 89 | |
XM_017003994.1 | XP_016859483.1 | 13 - 14 | 1554 | 89 | |
XM_017003999.1 | XP_016859488.1 | 13 - 14 | 1612 | 89 | |
XM_017004001.1 | XP_016859490.1 | 13 - 14 | 1554 | 89 | |
XM_017004002.1 | XP_016859491.1 | 13 - 14 | 1472 | 89 | |
GenBank mRNA | BC028361.2 | - | 13 - 14 | 1559 | 89 |