Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location
Chr.17: 7884764 - 7912757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005271.2 NP_001005271.2 28 - 29 4539 92
NM_001005273.2 NP_001005273.1 28 - 29 4510 92
NM_005852.3 NP_005843.2 28 - 29 4510 92
XM_005256427.4 XP_005256484.1 28 - 29 4580 92
XM_005256428.4 XP_005256485.1 28 - 29 4580 92
XM_005256429.4 XP_005256486.1 28 - 29 4580 92
XM_006721423.3 XP_006721486.1 28 - 29 4580 92
XM_006721424.3 XP_006721487.1 28 - 29 4555 92
XM_006721428.3 XP_006721491.1 28 - 29 4580 92
XM_017024061.1 XP_016879550.1 28 - 29 4562 92
XM_017024062.1 XP_016879551.1 28 - 29 4537 92
XM_017024063.1 XP_016879552.1 28 - 29 4495 92
XM_017024064.1 XP_016879553.1 28 - 29 4555 92
XM_017024065.1 XP_016879554.1 27 - 28 4376 92
XM_017024066.1 XP_016879555.1 29 - 30 4315 92
XM_017024067.1 XP_016879556.1 29 - 30 4498 92
XM_017024068.1 XP_016879557.1 28 - 29 4471 92
XM_017024069.1 XP_016879558.1 28 - 29 4362 92
XM_017024070.1 XP_016879559.1 28 - 29 4537 92
GenBank mRNA AK125928.1 - 5 - 6 923 92
AK296937.1 - 10 - 11 1304 92
AK308362.1 - 6 - 7 670 92
BC110648.1 - 4 - 5 343 92
U91543.1 - 28 - 29 4510 92

View Full Product Details