Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location
Chr.17: 7884764 - 7912757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005271.2 NP_001005271.2 31 - 32 4970 83
NM_001005273.2 NP_001005273.1 31 - 32 4941 83
NM_005852.3 NP_005843.2 31 - 32 4941 83
XM_005256427.4 XP_005256484.1 31 - 32 5011 83
XM_005256428.4 XP_005256485.1 31 - 32 5011 83
XM_005256429.4 XP_005256486.1 31 - 32 5011 83
XM_006721423.3 XP_006721486.1 31 - 32 5011 83
XM_006721424.3 XP_006721487.1 31 - 32 4986 83
XM_006721428.3 XP_006721491.1 31 - 32 5011 83
XM_017024061.1 XP_016879550.1 31 - 32 4993 83
XM_017024062.1 XP_016879551.1 31 - 32 4968 83
XM_017024063.1 XP_016879552.1 31 - 32 4926 83
XM_017024064.1 XP_016879553.1 31 - 32 4986 83
XM_017024065.1 XP_016879554.1 30 - 31 4807 83
XM_017024066.1 XP_016879555.1 32 - 33 4746 83
XM_017024067.1 XP_016879556.1 32 - 33 4929 83
XM_017024068.1 XP_016879557.1 31 - 32 4902 83
XM_017024069.1 XP_016879558.1 31 - 32 4793 83
XM_017024070.1 XP_016879559.1 31 - 32 4968 83
GenBank mRNA AK125928.1 - 8 - 9 1354 83
AK296725.1 - 6 - 7 808 83
AK296937.1 - 13 - 14 1735 83
AK308362.1 - 9 - 10 1101 83
BC110648.1 - 7 - 8 774 83
U91543.1 - 31 - 32 4941 83

View Full Product Details