Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location
Chr.17: 7884764 - 7912757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005271.2 NP_001005271.2 35 - 36 5436 68
NM_001005273.2 NP_001005273.1 35 - 36 5407 68
NM_005852.3 NP_005843.2 34 - 35 5305 68
XM_005256427.4 XP_005256484.1 35 - 36 5480 68
XM_005256428.4 XP_005256485.1 34 - 35 5375 68
XM_005256429.4 XP_005256486.1 35 - 36 5480 68
XM_006721423.3 XP_006721486.1 35 - 36 5477 68
XM_006721424.3 XP_006721487.1 35 - 36 5455 68
XM_006721428.3 XP_006721491.1 34 - 35 5375 68
XM_017024061.1 XP_016879550.1 35 - 36 5462 68
XM_017024062.1 XP_016879551.1 35 - 36 5437 68
XM_017024063.1 XP_016879552.1 35 - 36 5395 68
XM_017024064.1 XP_016879553.1 35 - 36 5452 68
XM_017024065.1 XP_016879554.1 34 - 35 5276 68
XM_017024066.1 XP_016879555.1 36 - 37 5215 68
XM_017024067.1 XP_016879556.1 36 - 37 5398 68
XM_017024068.1 XP_016879557.1 35 - 36 5371 68
XM_017024069.1 XP_016879558.1 35 - 36 5262 68
XM_017024070.1 XP_016879559.1 35 - 36 5437 68
GenBank mRNA AK125928.1 - 12 - 13 1823 68
AK296725.1 - 10 - 11 1274 68
AK308362.1 - 12 - 13 1465 68
BC110648.1 - 11 - 12 1240 68
U91543.1 - 35 - 36 5407 68

View Full Product Details