Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location
Chr.17: 7884764 - 7912757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005271.2 NP_001005271.2 39 - 40 6062 69
NM_001005273.2 NP_001005273.1 39 - 40 6033 69
NM_005852.3 NP_005843.2 38 - 39 5931 69
XM_005256427.4 XP_005256484.1 39 - 40 6152 69
XM_005256428.4 XP_005256485.1 38 - 39 6047 69
XM_005256429.4 XP_005256486.1 39 - 40 6106 69
XM_006721423.3 XP_006721486.1 39 - 40 6149 69
XM_006721424.3 XP_006721487.1 39 - 40 6127 69
XM_006721428.3 XP_006721491.1 38 - 39 6001 69
XM_017024061.1 XP_016879550.1 39 - 40 6134 69
XM_017024062.1 XP_016879551.1 39 - 40 6109 69
XM_017024063.1 XP_016879552.1 39 - 40 6067 69
XM_017024064.1 XP_016879553.1 39 - 40 6078 69
XM_017024065.1 XP_016879554.1 38 - 39 5948 69
XM_017024066.1 XP_016879555.1 40 - 41 5887 69
XM_017024067.1 XP_016879556.1 40 - 41 6070 69
XM_017024068.1 XP_016879557.1 39 - 40 6043 69
XM_017024069.1 XP_016879558.1 39 - 40 5934 69
XM_017024070.1 XP_016879559.1 39 - 40 6063 69
GenBank mRNA AK096555.1 - 2 - 3 1220 69
AK125928.1 - 16 - 17 2449 69
AK296937.1 - 21 - 22 2827 69
BC110648.1 - 15 - 16 1866 69
U91543.1 - 39 - 40 6033 69

View Full Product Details