Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
Chromosome Location
Chr.17: 7884764 - 7912757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005271.2 NP_001005271.2 5 - 6 978 59
NM_001005273.2 NP_001005273.1 5 - 6 949 59
NM_005852.3 NP_005843.2 5 - 6 949 59
XM_005256427.4 XP_005256484.1 5 - 6 1019 59
XM_005256428.4 XP_005256485.1 5 - 6 1019 59
XM_005256429.4 XP_005256486.1 5 - 6 1019 59
XM_005256431.4 XP_005256488.1 5 - 6 1019 59
XM_006721423.3 XP_006721486.1 5 - 6 1019 59
XM_006721424.3 XP_006721487.1 5 - 6 1006 59
XM_006721428.3 XP_006721491.1 5 - 6 1019 59
XM_017024061.1 XP_016879550.1 5 - 6 1019 59
XM_017024062.1 XP_016879551.1 5 - 6 1006 59
XM_017024063.1 XP_016879552.1 5 - 6 934 59
XM_017024064.1 XP_016879553.1 5 - 6 1006 59
XM_017024065.1 XP_016879554.1 5 - 6 1019 59
XM_017024066.1 XP_016879555.1 6 - 7 754 59
XM_017024067.1 XP_016879556.1 6 - 7 937 59
XM_017024068.1 XP_016879557.1 5 - 6 910 59
XM_017024069.1 XP_016879558.1 5 - 6 801 59
XM_017024070.1 XP_016879559.1 5 - 6 1006 59
GenBank mRNA U08379.1 - 2 - 3 440 59
U91543.1 - 5 - 6 949 59

View Full Product Details