The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003278.2 | NP_003269.2 | 1 - 2 | 200 | 85 |
GenBank mRNA | AK312093.1 | - | 1 - 2 | 208 | 85 |
BC011024.1 | - | 1 - 2 | 165 | 85 | |
BI837909.1 | - | 1 - 2 | 284 | 85 | |
CD616729.1 | - | 1 - 2 | 174 | 85 | |
CR541981.1 | - | 1 - 2 | 104 | 85 | |
CR542009.1 | - | 1 - 2 | 104 | 85 | |
X64559.1 | - | 1 - 2 | 197 | 85 |