Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MATN2
Entrez Gene ID
4147
Gene Name
matrilin 2
Gene Aliases
-
Chromosome Location
Chr.8: 97869021 - 98036720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317748.1 NP_001304677.1 16 - 17 2886 111
NM_002380.4 NP_002371.3 17 - 18 3009 111
NM_030583.3 NP_085072.2 17 - 18 2952 111
XM_005250920.1 XP_005250977.1 16 - 17 2560 111
XM_017013417.1 XP_016868906.1 16 - 17 2794 111
XM_017013418.1 XP_016868907.1 16 - 17 2503 111
GenBank mRNA AK027775.1 - 11 - 12 1640 111
AK094101.1 - - 1682 111
AK289721.1 - 17 - 18 3133 111
AL137638.1 - 15 - 16 2071 111
AL833931.1 - 17 - 18 2897 111
AY358895.1 - 16 - 17 2796 111
BC010444.1 - 17 - 18 2881 111
BC016394.1 - 14 - 15 1994 111
U69263.2 - 16 - 17 2841 111

View Full Product Details