Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MATN2
Entrez Gene ID
4147
Gene Name
matrilin 2
Gene Aliases
-
Chromosome Location
Chr.8: 97869021 - 98036720 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317748.1 NP_001304677.1 8 - 9 1624 75
NM_002380.4 NP_002371.3 9 - 10 1747 75
NM_030583.3 NP_085072.2 9 - 10 1747 75
XM_005250920.1 XP_005250977.1 9 - 10 1712 75
XM_017013417.1 XP_016868906.1 8 - 9 1589 75
XM_017013418.1 XP_016868907.1 9 - 10 1712 75
GenBank mRNA AK289721.1 - 9 - 10 1928 75
AL137638.1 - 7 - 8 809 75
AL833931.1 - 9 - 10 1635 75
AY358895.1 - 8 - 9 1534 75
BC010444.1 - 9 - 10 1676 75
BC016394.1 - 6 - 7 732 75
BX649093.1 - 6 - 7 801 75
U69263.2 - 8 - 9 1579 75

View Full Product Details