Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NHEJ1
Entrez Gene ID
79840
Gene Name
non-homologous end joining factor 1
Gene Aliases
XLF
Chromosome Location
Chr.2: 219075324 - 219160865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024782.2 NP_079058.1 4 - 5 672 116
GenBank mRNA AJ972687.1 - 4 - 5 616 116
AK022672.1 - 4 - 5 608 116
BC012732.2 - 4 - 5 593 116
BC030986.1 - 4 - 5 625 116
CR457291.1 - 3 - 4 526 116

View Full Product Details