Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NHEJ1
Entrez Gene ID
79840
Gene Name
non-homologous end joining factor 1
Gene Aliases
XLF
Chromosome Location
Chr.2: 219075324 - 219160865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024782.2 NP_079058.1 6 - 7 855 73
GenBank mRNA AJ972687.1 - 6 - 7 799 73
AK022672.1 - 6 - 7 791 73
BC008210.2 - 2 - 3 181 73
BC012732.2 - 6 - 7 776 73
BC030986.1 - 6 - 7 808 73
CR457291.1 - 5 - 6 709 73

View Full Product Details