The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130800.2 | NP_570712.1 | 1 - 2 | 47 | 79 |
XM_011545041.2 | XP_011543343.1 | 1 - 2 | 165 | 79 | |
XM_017017766.1 | XP_016873255.1 | 1 - 2 | 165 | 79 | |
XM_017017769.1 | XP_016873258.1 | 1 - 2 | 165 | 79 | |
GenBank mRNA | AJ297485.1 | - | 1 - 2 | 47 | 79 |
BC002544.2 | - | 1 - 2 | 42 | 79 |