The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080675.3 | NP_542406.2 | 7 - 8 | 520 | 71 |
XM_011528573.1 | XP_011526875.1 | 8 - 9 | 604 | 71 | |
XM_011528574.1 | XP_011526876.1 | 6 - 7 | 460 | 71 | |
XM_011528575.1 | XP_011526877.1 | 4 - 5 | 239 | 71 | |
XM_011528576.1 | XP_011526878.1 | 8 - 9 | 605 | 71 | |
GenBank mRNA | AF401350.1 | - | 7 - 8 | 520 | 71 |
BC026118.2 | - | 7 - 8 | 520 | 71 | |
BC029528.1 | - | 7 - 8 | 513 | 71 |