Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFM3
Entrez Gene ID
118427
Gene Name
olfactomedin 3
Gene Aliases
NOE3, NOELIN3, OPTIMEDIN
Chromosome Location
Chr.1: 101800028 - 101997234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288821.1 NP_001275750.1 3 - 4 538 77
NM_001288823.1 NP_001275752.1 4 - 5 850 77
NM_058170.3 NP_477518.2 3 - 4 789 77
NR_110210.1 4 - 5 850 77
NR_110211.1 3 - 4 789 77
NR_110212.1 3 - 4 538 77
XM_017000240.1 XP_016855729.1 4 - 5 693 77
GenBank mRNA AF397392.2 - 3 - 4 431 77
AF397393.2 - 3 - 4 431 77
AF397394.1 - 3 - 4 460 77
AF397395.1 - 3 - 4 460 77
AF397396.1 - 4 - 5 521 77
AF397397.1 - 4 - 5 521 77
AK095724.1 - 3 - 4 585 77
AK300870.1 - 3 - 4 538 77
AY358722.1 - 3 - 4 502 77
BC022531.1 - 3 - 4 547 77

View Full Product Details