The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005715.2 | NP_005706.1 | 7 - 8 | 1230 | 63 |
GenBank mRNA | AB020316.1 | - | 7 - 8 | 1037 | 63 |
AK292922.1 | - | 7 - 8 | 978 | 63 | |
AK315320.1 | - | 7 - 8 | 938 | 63 | |
BC093668.1 | - | 7 - 8 | 982 | 63 | |
BC093694.1 | - | 7 - 8 | 982 | 63 |