The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018368.3 | NP_060838.3 | 8 - 9 | 1043 | 114 |
XM_011535941.1 | XP_011534243.1 | 8 - 9 | 893 | 114 | |
GenBank mRNA | AF113224.1 | - | 8 - 9 | 763 | 114 |
AF208863.1 | - | 7 - 8 | 891 | 114 | |
AF211480.1 | - | 5 - 6 | 381 | 114 | |
AK002102.1 | - | 8 - 9 | 792 | 114 | |
AK225187.1 | - | 8 - 9 | 792 | 114 | |
AK225537.1 | - | 8 - 9 | 777 | 114 | |
AY136817.1 | - | 7 - 8 | 548 | 114 | |
BC010360.1 | - | 6 - 7 | 609 | 114 | |
BC047073.1 | - | 8 - 9 | 882 | 114 | |
BX647737.1 | - | 7 - 8 | 635 | 114 |