The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004370.5 | NP_004361.3 | 1 - 2 | 130 | 64 |
NM_080645.2 | NP_542376.2 | 1 - 2 | 130 | 64 | |
XM_011535434.1 | XP_011533736.1 | 1 - 2 | 274 | 64 | |
XM_011535435.1 | XP_011533737.1 | 1 - 2 | 274 | 64 | |
XM_011535436.2 | XP_011533738.1 | 1 - 2 | 485 | 64 | |
GenBank mRNA | AK307558.1 | - | 1 - 2 | 274 | 64 |
BC146868.1 | - | 1 - 2 | 74 | 64 | |
DA661773.1 | - | 1 - 2 | 274 | 64 | |
U73778.1 | - | 1 - 2 | 77 | 64 |