Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
Chromosome Location
Chr.6: 75084326 - 75206262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004370.5 NP_004361.3 19 - 20 4007 65
NM_080645.2 NP_542376.2 4 - 5 515 65
XM_011535434.1 XP_011533736.1 19 - 20 4151 65
XM_011535435.1 XP_011533737.1 18 - 19 3878 65
XM_011535436.2 XP_011533738.1 4 - 5 870 65
XM_017010252.1 XP_016865741.1 18 - 19 4403 65
GenBank mRNA BC146868.1 - 4 - 5 459 65
U73778.1 - 19 - 20 3954 65
U73779.1 - 3 - 4 385 65

View Full Product Details