Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
Chromosome Location
Chr.6: 75084326 - 75206262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004370.5 NP_004361.3 34 - 35 5966 84
NM_080645.2 NP_542376.2 19 - 20 2474 84
XM_011535434.1 XP_011533736.1 34 - 35 6110 84
XM_011535435.1 XP_011533737.1 33 - 34 5837 84
XM_011535436.2 XP_011533738.1 19 - 20 2829 84
XM_017010252.1 XP_016865741.1 33 - 34 6362 84
GenBank mRNA BC146868.1 - 19 - 20 2418 84

View Full Product Details