Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
Chromosome Location
Chr.6: 75084326 - 75206262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004370.5 NP_004361.3 43 - 44 7112 115
NM_080645.2 NP_542376.2 28 - 29 3620 115
XM_011535434.1 XP_011533736.1 43 - 44 7256 115
XM_011535435.1 XP_011533737.1 42 - 43 6983 115
XM_011535436.2 XP_011533738.1 28 - 29 3975 115
XM_017010252.1 XP_016865741.1 42 - 43 7508 115
GenBank mRNA BC146868.1 - 28 - 29 3564 115
U73778.1 - 43 - 44 7059 115

View Full Product Details