Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL12A1
Entrez Gene ID
1303
Gene Name
collagen type XII alpha 1 chain
Gene Aliases
BA209D8.1, BTHLM2, COL12A1L, DJ234P15.1, UCMD2
Chromosome Location
Chr.6: 75084326 - 75206262 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004370.5 NP_004361.3 61 - 62 8846 72
NM_080645.2 NP_542376.2 46 - 47 5354 72
XM_011535434.1 XP_011533736.1 61 - 62 8990 72
XM_011535435.1 XP_011533737.1 60 - 61 8717 72
XM_011535436.2 XP_011533738.1 46 - 47 5709 72
XM_017010252.1 XP_016865741.1 60 - 61 9242 72
GenBank mRNA AJ420501.1 - 6 - 7 333 72
BC146868.1 - 46 - 47 5298 72
U73778.1 - 61 - 62 8793 72

View Full Product Details