Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRHL2
Entrez Gene ID
79977
Gene Name
grainyhead like transcription factor 2
Gene Aliases
BOM, DFNA28, ECTDS, TFCP2L3
Chromosome Location
Chr.8: 101492440 - 101678105 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024915.3 NP_079191.2 15 - 16 2096 84
XM_011517305.2 XP_011515607.1 15 - 16 2127 84
XM_011517306.2 XP_011515608.1 15 - 16 1864 84
GenBank mRNA AK002034.1 - 2 - 3 120 84
AK023844.1 - 15 - 16 1778 84
AK292853.1 - 15 - 16 1778 84
BC069618.1 - 15 - 16 1778 84
BC069633.1 - 15 - 16 1838 84
BC069638.1 - 15 - 16 1778 84
BC129822.1 - 15 - 16 1778 84
BC129823.1 - 15 - 16 1778 84

View Full Product Details