The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182924.3 | NP_891554.1 | 7 - 8 | 1862 | 101 |
XM_011515520.2 | XP_011513822.1 | 7 - 8 | 1886 | 101 | |
XM_011515522.2 | XP_011513824.1 | 7 - 8 | 1886 | 101 | |
XM_011515523.2 | XP_011513825.1 | 7 - 8 | 1892 | 101 | |
XM_017012619.1 | XP_016868108.1 | 7 - 8 | 1886 | 101 | |
GenBank mRNA | AK027124.1 | - | 3 - 4 | 1226 | 101 |
AL833704.2 | - | 6 - 7 | 1820 | 101 | |
BC037988.1 | - | 7 - 8 | 1849 | 101 |