The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182924.3 | NP_891554.1 | 9 - 10 | 2115 | 132 |
XM_011515520.2 | XP_011513822.1 | 9 - 10 | 2139 | 132 | |
XM_011515522.2 | XP_011513824.1 | 9 - 10 | 2139 | 132 | |
XM_011515523.2 | XP_011513825.1 | 9 - 10 | 2145 | 132 | |
XM_011515524.2 | XP_011513826.1 | 3 - 4 | 339 | 132 | |
GenBank mRNA | AK027124.1 | - | 5 - 6 | 1479 | 132 |
AK074068.1 | - | 8 - 9 | 3516 | 132 | |
AK126808.1 | - | 2 - 3 | 1097 | 132 | |
BC037988.1 | - | 9 - 10 | 2102 | 132 |