The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168647.1 | NP_001162118.1 | 14 - 15 | 2215 | 145 |
NM_001168648.1 | NP_001162119.1 | 15 - 16 | 2305 | 145 | |
NM_001168649.1 | NP_001162120.1 | 14 - 15 | 2158 | 145 | |
NM_014927.3 | NP_055742.2 | 15 - 16 | 2305 | 145 | |
XM_011545471.2 | XP_011543773.1 | 14 - 15 | 2298 | 145 | |
XM_011545472.2 | XP_011543774.1 | 14 - 15 | 2162 | 145 | |
XM_017029358.1 | XP_016884847.1 | 13 - 14 | 2072 | 145 | |
GenBank mRNA | AB020709.1 | - | 15 - 16 | 2281 | 145 |
AF418269.1 | - | 15 - 16 | 1861 | 145 | |
AF418270.1 | - | 15 - 16 | 1861 | 145 | |
AK056549.1 | - | 4 - 5 | 657 | 145 | |
AK294728.1 | - | 14 - 15 | 2120 | 145 | |
BC126121.1 | - | 15 - 16 | 1860 | 145 | |
BC136289.1 | - | 15 - 16 | 1860 | 145 | |
BC143839.1 | - | 14 - 15 | 1770 | 145 |