Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XYLT1
Entrez Gene ID
64131
Gene Name
xylosyltransferase 1
Gene Aliases
DBQD2, PXYLT1, XT-I, XT1, XTI, XYLTI, xylT-I
Chromosome Location
Chr.16: 17102324 - 17470934 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022166.3 NP_071449.1 9 - 10 2114 103
XM_017023539.1 XP_016879028.1 9 - 10 2167 103
XM_017023540.1 XP_016879029.1 9 - 10 2167 103
GenBank mRNA AJ277441.1 - 7 - 8 1633 103
AJ539163.1 - 9 - 10 2114 103

View Full Product Details