Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
Chromosome Location
Chr.7: 87345635 - 87399796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143935.1 NP_001137407.1 12 - 13 1391 146
NM_021151.3 NP_066974.2 11 - 12 1307 146
XM_011516337.2 XP_011514639.1 11 - 12 1307 146
XM_017012370.1 XP_016867859.1 12 - 13 1381 146
GenBank mRNA AF073770.1 - 11 - 12 1198 146
AF168793.1 - 11 - 12 1198 146
AK127901.1 - 12 - 13 1347 146
AK300114.1 - 6 - 7 637 146
AK313487.1 - 11 - 12 1250 146
BC039004.1 - 11 - 12 1247 146

View Full Product Details