The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001925.2 | NP_001916.1 | 1 - 2 | 110 | 95 |
GenBank mRNA | BC093959.1 | - | 1 - 2 | 34 | 95 |
BC112091.1 | - | 1 - 2 | 34 | 95 | |
BU616655.1 | - | 2 - 3 | 95 | ||
DN998607.1 | - | 1 - 2 | 59 | 95 | |
X65977.1 | - | 1 - 2 | 36 | 95 |