The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033117.2 | NP_001028289.1 | 3 - 4 | 1162 | 67 |
NM_014850.3 | NP_055665.1 | 3 - 4 | 1162 | 67 | |
XM_011534295.1 | XP_011532597.1 | 3 - 4 | 1162 | 67 | |
XM_011534296.1 | XP_011532598.1 | 4 - 5 | 1279 | 67 | |
XM_017007574.1 | XP_016863063.1 | 3 - 4 | 1500 | 67 | |
XM_017007575.1 | XP_016863064.1 | 3 - 4 | 1500 | 67 | |
XM_017007576.1 | XP_016863065.1 | 5 - 6 | 669 | 67 | |
XM_017007577.1 | XP_016863066.1 | 5 - 6 | 608 | 67 | |
XM_017007578.1 | XP_016863067.1 | 3 - 4 | 1528 | 67 | |
XM_017007579.1 | XP_016863068.1 | 3 - 4 | 1496 | 67 | |
XM_017007580.1 | XP_016863069.1 | 5 - 6 | 729 | 67 | |
GenBank mRNA | AB007871.2 | - | 3 - 4 | 386 | 67 |
AF427144.1 | - | 3 - 4 | 530 | 67 | |
AF464189.1 | - | 3 - 4 | 690 | 67 | |
AK094355.1 | - | 3 - 4 | 468 | 67 | |
BC039300.1 | - | 3 - 4 | 497 | 67 |