Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA1A
Entrez Gene ID
51365
Gene Name
phospholipase A1 member A
Gene Aliases
PS-PLA1, PSPLA1
Chromosome Location
Chr.3: 119597848 - 119629811 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206960.1 NP_001193889.1 1 - 2 146 77
NM_015900.3 NP_056984.1 1 - 2 146 77
NR_120610.1 1 - 2 146 77
GenBank mRNA AF035268.1 - 1 - 2 111 77
AF035269.1 - 1 - 2 111 77
AK222705.1 - 1 - 2 109 77
AK313519.1 - 1 - 2 109 77
BC035347.1 - 1 - 2 98 77
BC047703.2 - 1 - 2 98 77
DB189568.1 - 1 - 2 147 77
DB255030.1 - 1 - 2 119 78
U37591.1 - 1 - 2 106 77

View Full Product Details