Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA1A
Entrez Gene ID
51365
Gene Name
phospholipase A1 member A
Gene Aliases
PS-PLA1, PSPLA1
Chromosome Location
Chr.3: 119597848 - 119629811 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206960.1 NP_001193889.1 6 - 7 766 79
NM_001206961.1 NP_001193890.1 5 - 6 612 79
NM_001293225.1 NP_001280154.1 6 - 7 809 79
NM_015900.3 NP_056984.1 6 - 7 814 79
NR_120610.1 6 - 7 814 79
XM_017006572.1 XP_016862061.1 7 - 8 846 79
GenBank mRNA AF035268.1 - 6 - 7 779 79
AF035269.1 - 6 - 7 779 79
AK222705.1 - 6 - 7 777 79
AK301880.1 - 5 - 6 585 79
AK313519.1 - 6 - 7 777 79
BC035347.1 - 6 - 7 766 79
BC047703.2 - 6 - 7 718 79
BX647516.1 - 6 - 7 809 79
U37591.1 - 6 - 7 774 79

View Full Product Details