Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
Chromosome Location
Chr.1: 28887091 - 29120046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166005.1 NP_001159477.1 9 - 10 1493 103
NM_001166006.1 NP_001159478.1 9 - 10 1566 103
NM_001166007.1 NP_001159479.1 9 - 10 1476 103
NM_004437.3 NP_004428.1 10 - 11 1556 103
NM_203342.2 NP_976217.1 10 - 11 1573 103
NM_203343.2 NP_976218.1 8 - 9 1388 103
XM_005245753.1 XP_005245810.1 9 - 10 1568 103
XM_005245757.1 XP_005245814.1 9 - 10 1568 103
XM_005245760.1 XP_005245817.1 9 - 10 1568 103
XM_005245761.1 XP_005245818.1 9 - 10 1568 103
XM_005245763.1 XP_005245820.1 9 - 10 1568 103
XM_005245764.1 XP_005245821.1 9 - 10 1568 103
XM_005245765.1 XP_005245822.1 9 - 10 1568 103
XM_005245768.1 XP_005245825.1 9 - 10 1568 103
XM_005245769.1 XP_005245826.1 9 - 10 1568 103
XM_005245770.1 XP_005245827.1 9 - 10 1568 103
XM_005245772.4 XP_005245829.1 9 - 10 1568 103
XM_005245773.4 XP_005245830.1 9 - 10 1568 103
XM_005245774.1 XP_005245831.1 9 - 10 1568 103
XM_006710434.1 XP_006710497.1 9 - 10 1568 103
XM_006710439.1 XP_006710502.1 9 - 10 1568 103
XM_011540956.1 XP_011539258.1 9 - 10 1568 103
XM_011540957.1 XP_011539259.1 9 - 10 1568 103
XM_011540958.1 XP_011539260.1 9 - 10 1568 103
XM_011540959.1 XP_011539261.1 9 - 10 1568 103
XM_011540960.1 XP_011539262.1 9 - 10 1568 103
XM_011540961.1 XP_011539263.1 9 - 10 1568 103
XM_011540962.1 XP_011539264.1 9 - 10 1568 103
XM_011540963.2 XP_011539265.1 9 - 10 1568 103
XM_011540964.1 XP_011539266.1 9 - 10 1464 103
XM_011540965.2 XP_011539267.1 9 - 10 1568 103
XM_017000581.1 XP_016856070.1 8 - 9 1463 103
XM_017000582.1 XP_016856071.1 8 - 9 1463 103
XM_017000583.1 XP_016856072.1 9 - 10 1568 103
XM_017000584.1 XP_016856073.1 9 - 10 1859 103
XM_017000585.1 XP_016856074.1 9 - 10 1856 103
XM_017000586.1 XP_016856075.1 9 - 10 1568 103
XM_017000587.1 XP_016856076.1 9 - 10 1568 103
XM_017000588.1 XP_016856077.1 9 - 10 1568 103
XM_017000589.1 XP_016856078.1 8 - 9 1463 103
XM_017000590.1 XP_016856079.1 8 - 9 1463 103
XM_017000591.1 XP_016856080.1 9 - 10 1568 103
XM_017000592.1 XP_016856081.1 9 - 10 1568 103
XM_017000593.1 XP_016856082.1 9 - 10 1464 103
XM_017000594.1 XP_016856083.1 9 - 10 1464 103
XM_017000595.1 XP_016856084.1 9 - 10 1464 103
XM_017000596.1 XP_016856085.1 9 - 10 1464 103
XM_017000597.1 XP_016856086.1 9 - 10 1464 103
XM_017000598.1 XP_016856087.1 9 - 10 1464 103
XM_017000599.1 XP_016856088.1 9 - 10 1464 103
XM_017000600.1 XP_016856089.1 9 - 10 1464 103
XM_017000601.1 XP_016856090.1 9 - 10 1464 103
XM_017000602.1 XP_016856091.1 9 - 10 1464 103
XM_017000603.1 XP_016856092.1 9 - 10 1464 103
XM_017000604.1 XP_016856093.1 9 - 10 1464 103
GenBank mRNA AB209649.1 - 9 - 10 1476 103
AF156225.1 - 8 - 9 1366 103
AK310655.1 - 8 - 9 1404 103
BC039079.1 - 9 - 10 1563 103
BC096104.1 - 6 - 7 782 103
BC096105.1 - 8 - 9 1100 103
BC113841.1 - 8 - 9 1100 103
BC114450.1 - 8 - 9 1100 103
J03796.1 - 8 - 9 1307 103
M14993.1 - 10 - 11 1537 103
M61733.1 - 10 - 11 1554 103

View Full Product Details