Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
Chromosome Location
Chr.1: 28887091 - 29120046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004437.3 NP_004428.1 2 - 3 579 145
NM_203342.2 NP_976217.1 2 - 3 596 145

View Full Product Details