Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
Chromosome Location
Chr.1: 28887091 - 29120046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166005.1 NP_001159477.1 5 - 6 956 90
NM_001166006.1 NP_001159478.1 5 - 6 1029 90
NM_001166007.1 NP_001159479.1 5 - 6 939 90
NM_004437.3 NP_004428.1 6 - 7 1019 90
NM_203342.2 NP_976217.1 6 - 7 1036 90
NM_203343.2 NP_976218.1 4 - 5 851 90
XM_005245753.1 XP_005245810.1 5 - 6 1031 90
XM_005245757.1 XP_005245814.1 5 - 6 1031 90
XM_005245760.1 XP_005245817.1 5 - 6 1031 90
XM_005245761.1 XP_005245818.1 5 - 6 1031 90
XM_005245763.1 XP_005245820.1 5 - 6 1031 90
XM_005245764.1 XP_005245821.1 5 - 6 1031 90
XM_005245765.1 XP_005245822.1 5 - 6 1031 90
XM_005245768.1 XP_005245825.1 5 - 6 1031 90
XM_005245769.1 XP_005245826.1 5 - 6 1031 90
XM_005245770.1 XP_005245827.1 5 - 6 1031 90
XM_005245772.4 XP_005245829.1 5 - 6 1031 90
XM_005245773.4 XP_005245830.1 5 - 6 1031 90
XM_005245774.1 XP_005245831.1 5 - 6 1031 90
XM_006710434.1 XP_006710497.1 5 - 6 1031 90
XM_006710439.1 XP_006710502.1 5 - 6 1031 90
XM_011540956.1 XP_011539258.1 5 - 6 1031 90
XM_011540957.1 XP_011539259.1 5 - 6 1031 90
XM_011540958.1 XP_011539260.1 5 - 6 1031 90
XM_011540959.1 XP_011539261.1 5 - 6 1031 90
XM_011540960.1 XP_011539262.1 5 - 6 1031 90
XM_011540961.1 XP_011539263.1 5 - 6 1031 90
XM_011540962.1 XP_011539264.1 5 - 6 1031 90
XM_011540963.2 XP_011539265.1 5 - 6 1031 90
XM_011540964.1 XP_011539266.1 5 - 6 927 90
XM_011540965.2 XP_011539267.1 5 - 6 1031 90
XM_017000581.1 XP_016856070.1 4 - 5 926 90
XM_017000582.1 XP_016856071.1 4 - 5 926 90
XM_017000583.1 XP_016856072.1 5 - 6 1031 90
XM_017000584.1 XP_016856073.1 5 - 6 1322 90
XM_017000585.1 XP_016856074.1 5 - 6 1319 90
XM_017000586.1 XP_016856075.1 5 - 6 1031 90
XM_017000587.1 XP_016856076.1 5 - 6 1031 90
XM_017000588.1 XP_016856077.1 5 - 6 1031 90
XM_017000589.1 XP_016856078.1 4 - 5 926 90
XM_017000590.1 XP_016856079.1 4 - 5 926 90
XM_017000591.1 XP_016856080.1 5 - 6 1031 90
XM_017000592.1 XP_016856081.1 5 - 6 1031 90
XM_017000593.1 XP_016856082.1 5 - 6 927 90
XM_017000594.1 XP_016856083.1 5 - 6 927 90
XM_017000595.1 XP_016856084.1 5 - 6 927 90
XM_017000596.1 XP_016856085.1 5 - 6 927 90
XM_017000597.1 XP_016856086.1 5 - 6 927 90
XM_017000598.1 XP_016856087.1 5 - 6 927 90
XM_017000599.1 XP_016856088.1 5 - 6 927 90
XM_017000600.1 XP_016856089.1 5 - 6 927 90
XM_017000601.1 XP_016856090.1 5 - 6 927 90
XM_017000602.1 XP_016856091.1 5 - 6 927 90
XM_017000603.1 XP_016856092.1 5 - 6 927 90
XM_017000604.1 XP_016856093.1 5 - 6 927 90
GenBank mRNA AB209649.1 - 5 - 6 939 90
AF156225.1 - 4 - 5 829 90
AK310655.1 - 4 - 5 867 90
BC039079.1 - 5 - 6 1026 90
BC096104.1 - 2 - 3 245 90
BC096105.1 - 4 - 5 563 90
BC113841.1 - 4 - 5 563 90
BC114450.1 - 4 - 5 563 90
J03796.1 - 4 - 5 770 90
M14993.1 - 6 - 7 1000 90
M61733.1 - 6 - 7 1017 90

View Full Product Details