The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000223.3 | NP_000214.1 | 1 - 2 | 584 | 91 |
GenBank mRNA | AK313747.1 | - | 1 - 2 | 598 | 91 |
D78367.1 | - | 1 - 2 | 584 | 91 |