The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004304.4 | NP_004295.2 | 18 - 19 | 4022 | 114 |
GenBank mRNA | AB209477.4 | - | 18 - 19 | 3559 | 114 |
EU788003.1 | - | 18 - 19 | 3977 | 114 | |
EU788004.1 | - | 18 - 19 | 3977 | 114 | |
EU788005.1 | - | 18 - 19 | 3977 | 114 | |
EU788006.1 | - | 18 - 19 | 3977 | 114 | |
EU788007.1 | - | 18 - 19 | 3977 | 114 | |
EU788008.1 | - | 18 - 19 | 3977 | 114 | |
EU788009.1 | - | 18 - 19 | 3977 | 114 | |
EU788010.1 | - | 18 - 19 | 3977 | 114 | |
FJ838786.1 | - | 18 - 19 | 3977 | 114 | |
FJ838787.1 | - | 18 - 19 | 3977 | 114 | |
GU128152.1 | - | 18 - 19 | 3070 | 114 | |
GU128153.1 | - | 18 - 19 | 3070 | 114 | |
GU128154.1 | - | 18 - 19 | 3070 | 114 | |
GU128155.1 | - | 18 - 19 | 3070 | 114 | |
GU128156.1 | - | 18 - 19 | 3070 | 114 | |
GU128157.1 | - | 18 - 19 | 3070 | 114 | |
U62540.1 | - | 18 - 19 | 3981 | 114 | |
U66559.1 | - | 18 - 19 | 3218 | 114 |