The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006815.3 | NP_006806.1 | 2 - 3 | 479 | 80 |
GenBank mRNA | AK298167.1 | - | 2 - 3 | 373 | 80 |
AK312975.1 | - | 2 - 3 | 454 | 80 | |
BC025957.1 | - | 2 - 3 | 394 | 80 | |
BI086507.1 | - | 2 - 3 | 407 | 80 | |
BP226125.1 | - | 2 - 3 | 478 | 80 | |
CR541663.1 | - | 2 - 3 | 371 | 80 | |
CR541682.1 | - | 2 - 3 | 371 | 80 | |
X92098.1 | - | 2 - 3 | 398 | 80 |