Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC10
Entrez Gene ID
7109
Gene Name
trafficking protein particle complex 10
Gene Aliases
EHOC-1, EHOC1, GT334, TMEM1, TRS130, TRS30
Chromosome Location
Chr.21: 44012325 - 44106552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003274.4 NP_003265.3 18 - 19 3040 83
XM_005261168.4 XP_005261225.1 18 - 19 2910 83
XM_011529714.2 XP_011528016.1 19 - 20 3144 83
XM_011529715.2 XP_011528017.1 19 - 20 3021 83
XM_011529716.2 XP_011528018.1 18 - 19 2997 83
XM_011529717.2 XP_011528019.1 19 - 20 3144 83
XM_011529718.2 XP_011528020.1 19 - 20 3144 83
XM_011529719.2 XP_011528021.1 17 - 18 2700 83
XM_011529720.2 XP_011528022.1 19 - 20 3144 83
XM_011529721.2 XP_011528023.1 16 - 17 2517 83
XM_011529722.2 XP_011528024.1 16 - 17 2483 83
XM_011529723.2 XP_011528025.1 15 - 16 2380 83
XM_011529727.2 XP_011528029.1 15 - 16 2385 83
XM_011529728.2 XP_011528030.1 16 - 17 2666 83
XM_011529729.2 XP_011528031.1 15 - 16 2290 83
XM_017028454.1 XP_016883943.1 17 - 18 2886 83
XM_017028455.1 XP_016883944.1 11 - 12 1886 83
GenBank mRNA AK295369.1 - 5 - 6 660 83
AK300912.1 - 2 - 3 198 83
AK303570.1 - 6 - 7 645 83
AL833668.1 - 1 - 2 1896 83
BC101728.1 - 18 - 19 2960 83
U19252.1 - 18 - 19 3008 83
U61500.1 - 18 - 19 3024 83

View Full Product Details