Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRP1
Entrez Gene ID
4035
Gene Name
LDL receptor related protein 1
Gene Aliases
A2MR, APOER, APR, CD91, IGFBP3R, LRP, LRP1A, TGFBR5
Chromosome Location
Chr.12: 57128401 - 57213377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002332.2 NP_002323.2 23 - 24 4263 81
XM_017019303.1 XP_016874792.1 24 - 25 4412 81
GenBank mRNA X13916.1 - 23 - 24 4263 81

View Full Product Details