The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002332.2 | NP_002323.2 | 51 - 52 | 8780 | 110 |
XM_017019303.1 | XP_016874792.1 | 52 - 53 | 8929 | 110 | |
GenBank mRNA | AB209498.1 | - | 29 - 30 | 4528 | 110 |
X13916.1 | - | 51 - 52 | 8780 | 110 |